Learn More
ATCC Escherichia coli; Strain P1
Supplier:  ATCC BAA1427
Encompass
Escherichia coli strain P1 is a whole-genome sequenced bacterium that was isolated from cattle hides. This strain is non-toxigenic and is recommended by the USDA as a surrogate indicator organism to measure changes in microbial counts.
Catalog No. 50-238-3018