Learn More
TARGETMOL CHEMICALS INC Escitalopram 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T018525MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Escitalopram (Seroplex) is a furancarbonitrile that is one of the SEROTONIN UPTAKE INHIBITORS used as an antidepressant. The drug is also effective in reducing ethanol uptake in alcoholics and is used in depressed patients who also suffer from tardive dyskinesia in preference to tricyclic antidepressants, which aggravate this condition. Purity 99.82%
Catalog No. 50-288-0710