Learn More
Selleck Chemical LLC Escitalopram Oxalate S4064-1g
SDP
Supplier:  Selleck Chemical LLC S40641G
Encompass_Preferred
Escitalopram Oxalate is a selective serotonin (5-HT) reuptake inhibitor (SSRI) with Ki of 0 89 nM
Catalog No. 50-310-8839