Learn More
Medchemexpress LLC Escitalopram (oxalate) | 219861-08-2 | 99.9% | 414.43 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY14258A
Encompass_Preferred
Escitalopram oxalate, the S-enantiomer of racemic Citalopram, is a selective serotonin reuptake inhibitor (SSRI). It exhibits approximately 30-fold higher binding affinity than its R(-)-enantiomer and demonstrates selectivity over both dopamine transporter (DAT) and norepinephrine transporter (NET). This compound is an antidepressant used in research for major depression, intended for research use only.
  • Selective serotonin reuptake inhibitor (SSRI)
  • Exhibits approximately 30-fold higher binding affinity than its R(-)-enantiomer
  • Demonstrates selectivity over dopamine and norepinephrine transporters
  • Ameliorates cognitive impairments and attenuates phosphorylated tau accumulation in stressed rats
  • Significantly reduces plaque load in APP-PS1 hemizygous female mice
Catalog No. 50-003-65334