Learn More
Medchemexpress LLC Escitalopram (oxalate) | 219861-08-2 | 99.9% | C22H23FN2O5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14258100MG
Encompass_Preferred
Escitalopram oxalate, the S-enantiomer of racemic Citalopram, is a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM for the serotonin transporter. It exhibits approximately 30-fold higher binding affinity than its R(-)-enantiomer and selectivity over dopamine and norepinephrine transporters. This compound is primarily utilized as an antidepressant in the research of major depression and for various neurological disease studies. This product is intended for research and analytical applications only.
  • High purity for reliable research applications
  • Selective serotonin reuptake inhibitor
  • Suitable for neurological disease research
  • Used in studies of major depression
  • For research use only
Catalog No. 50-003-55637