Learn More
Medchemexpress LLC Escitalopram oxalate | 219861-08-2 | 99.97% | C20H21FN2O.C2H2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14258A50MG
Encompass_Preferred
Escitalopram oxalate, the S-enantiomer of racemic Citalopram, is a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM. It exhibits approximately 30-fold higher binding affinity than its R(-)-enantiomer and demonstrates selectivity over both the dopamine transporter (DAT) and norepinephrine transporter (NET). It is an antidepressant used in the research of major depression.
  • Ameliorates cognitive impairments and selectively attenuates phosphorylated tau accumulation in stressed rats.
  • Significantly reduces plaque load in APP-PS1 hemizygous female mice.
Catalog No. 50-003-53578