Learn More
Medchemexpress LLC Escitalopram oxalate | 219861-08-2 | MFCD06407826 | 100.0% | C22H23FN2O5 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14258AR10MG
Encompass_Preferred
Escitalopram oxalate is the oxalate salt of escitalopram (the S-enantiomer of citalopram) supplied as a high-purity analytical standard for research and analytical applications. It is a selective serotonin reuptake inhibitor with high affinity for the serotonin transporter and is provided as a solid reference material for laboratory use only.
  • High purity analytical standard (99.98%).
  • CAS number 219861-08-2 for unambiguous identification.
  • Intended for research and analytical applications, not for human use.
  • Available in multiple small package sizes (5-500 mg) for method development.
  • Reported high affinity for the serotonin transporter (Ki ≈ 0.89 nM).
Catalog No. 50-002-61020