Learn More
Medchemexpress LLC Escitalopram (oxalate) (Standard) | 219861-08-2 | 99.7% | C22H23FN2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY14258A10MM/1ML
Encompass_Preferred
Escitalopram ((S)-Citalopram) oxalate is the S-enantiomer of racemic Citalopram, functioning as a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM. It exhibits significantly higher binding affinity compared to its R(-)-enantiomer and demonstrates selectivity over both dopamine and norepinephrine transporters. This antidepressant is utilized in research for major depression.
  • Selective serotonin reuptake inhibitor (SSRI)
  • Ki of 0.89 nM (serotonin transporter)
  • Approximately 30-fold higher binding affinity than its R(-)-enantiomer
  • Selective over dopamine transporter (DAT) and norepinephrine transporter (NET)
  • Used for major depression research
Catalog No. 50-003-55632