Learn More
Medchemexpress LLC Escitalopram oxalate (Standard) | 219861-08-2 | 99.98% | C22H23FN2O5 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1425850MG
Encompass_Preferred
Escitalopram oxalate (Standard) is an analytical standard for research and analytical applications. It is the S-enantiomer of racemic Citalopram, acting as a selective serotonin reuptake inhibitor (SSRI) with a Ki of 0.89 nM. It shows high binding affinity and selectivity over dopamine and norepinephrine transporters, making it useful in major depression research.
  • Analytical standard for research and analytical applications
  • Functions as a selective serotonin reuptake inhibitor (SSRI)
  • Used in qualitative, quantitative, and methodological research
  • Suitable for HPLC, GC, and MS
  • Antidepressant for major depression research
  • High binding affinity and selectivity over other transporters
Catalog No. 50-003-53681