Learn More
Enzo Life Sciences Esculetin (1g). CAS: 305-01-1
SDP
Supplier:  Enzo Life Sciences BMLEI1000001
Encompass_Preferred
Inhibitor of 5-lipoxygenase (ID50=4 µM) and 12-lipoxygenase (ID50=2.5 µM). Does not inhibit cyclooxygenase. Alternative name: 6,7-Dihydroxycoumarin. Purity: ≥98% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-0543