Learn More
Alkali Scientific Esculin, 1 Kilogram
SDP
Supplier:  Alkali Scientific ESC001CP
Encompass_Preferred
Esculin is a biochemical reagent supplied in a 1 kg bottle, typically used in laboratory settings. This chemical is widely utilized for tests involving the isolation and identification of microorganisms, particularly in the differentiation of certain Gram-positive bacteria. The compound is used in biochemical assays where it acts as a substrate in the determination of enzyme activities like esculin hydrolysis, which is a hallmark of certain bacterial species. It’s also used for preparing specific media for bacterial identification and fermentation tests, as well as in other microbiological procedures that require a substrate for enzyme activity. This high-purity reagent is perfect for academic research, quality control, and other laboratory analyses.
Catalog No. 50-006-60316