Learn More
Alkali Scientific Esculin, 25 G
SDP
Supplier:  Alkali Scientific ESC003CP
Encompass_Preferred
Esculin, a plant-derived glycoside, is commonly used in biochemical research. This 25g laboratory-grade quantity is utilized in a variety of laboratory tests, primarily for its ability to fluoresce under UV light once hydrolyzed by enzymes. It is most often employed in microbiological assays to identify enterococci, which break down esculin in the presence of bile salts. Additionally, esculin is used in enzyme activity studies, as well as in the development of reagents for chemical analysis. This high-quality reagent is essential for accurate and reliable scientific research.
Catalog No. 50-006-59939