Learn More
Uline ESD Conductive Shippers, Graphite, 6
Supplier:  Uline S12740CS
Encompass
Provides total Faraday cage protection to ship or store circuit boards, Static dissipative 200 lb test corrugated with graphite coating, No assembly required, MEETS REQUIREMENT OF EIA 541
Catalog No. NC2594638