Learn More
Electron Microscopy Sciences EMS ESD Tweezers Boley Style AA, Anti-Acid/Anti-MagSS, 130 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 78325AA
Encompass_Preferred
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed. Straight, sharp, fine tips.
OAL: 130 mm
Tips: Anti-Acid/Anti-MagSS
Material: Straight, sharp, fine
Catalog No. 50-193-0604