Learn More
Selleck Chemical LLC ESI-05 S3382-25mg
SDP
Supplier:  Selleck Chemical LLC S338225MG
Encompass_Preferred
ESI-05 (NSC 116966) is a specific EPAC2 (exchange protein directly activated by cAMP 2) antagonist with IC50 of 0 4 M ESI-05 (NSC 116966) inhibits cAMP-mediated activation of EPAC2 and EAPC2 mediated Rap1 activation
Catalog No. 50-310-9316