Learn More
Medchemexpress LLC ESI1 | 99.76% | 355.82 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY4351550MG
Encompass_Preferred
ESI1 is a small molecule epigenetic silencing inhibitor that triggers the formation of nuclear condensates of key lipid metabolism regulators SREBP1/2. This action concentrates transcriptional co-activators to drive lipid/cholesterol biosynthesis.
  • Promotes myelin regeneration in demyelinated animal models.
  • Facilitates de novo myelination on regenerating CNS axons.
  • Reverses age-related declines in cognitive abilities.
Catalog No. 50-004-03793