Learn More
Medchemexpress LLC Eslicarbazepine acetate | 236395-14-5 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB070350MG
Encompass_Preferred
Eslicarbazepine acetate (BIA 2-093) is an antiepileptic agent and a dual inhibitor of β-Secretase and voltage-gated sodium channels. It is a prodrug that is activated to eslicarbazepine (S-licarbazepine), which is an active metabolite of oxcarbazepine. Its mechanism of action is similar to oxcarbazepine, potentially offering improved tolerability.
  • Antiepileptic agent
  • Dual inhibitor of β-secretase
  • Dual inhibitor of voltage-gated sodium channels
  • Prodrug activated to eslicarbazepine (S-licarbazepine)
  • Active metabolite of oxcarbazepine
  • Similar mechanism of action to oxcarbazepine
Catalog No. 50-004-06851