Learn More
Cayman Chemical Esmolol AcIdhydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 352955 MG
Encompass_Preferred
An active metabolite of esmolol; inhibits isoproterenol-induced increases in the beating rate of isolated guinea pig right atria (pA2 = 3.73); prevents isoproterenol-induced tachycardia and hypotension in anesthetized dogs; a potential impurity in commercial preparations of esmolol
Catalog No. 50-290-9299