Learn More
Cayman Chemical EsmololhydrochlorIde 100mg
SDP
Supplier:  Cayman Chemical 22581100 MG
Encompass_Preferred
A β1-AR antagonist; binds to β1-ARs (Kd = 100 nM in isolated cardiac myocytes); 34-fold selective for β1- over β2-ARs; inhibits ICa.L and the INa in rat cardiac myocytes (IC50s = 50 and 169 μM, respectively)
Catalog No. 50-298-6270