Learn More
Cayman Chemical Esomeprazole 250mg
SDP
Supplier:  Cayman Chemical 40851250 MG
Encompass_Preferred
An H+/K+-ATPase inhibitor; reversibly inhibits the activity of the H+/K+-ATPase in an enzyme assay; inhibits histamine-induced gastric acid secretion in rats at 30 mg/kg, s.c.
Catalog No. 50-294-2529