Learn More
Apexbio Technology LLC Esomeprazole Magnesium 161973-10-0 50mg
SDP
Supplier:  Apexbio Technology LLC B220050
Encompass_Preferred
Esomeprazole Magnesium is a small-molecule inhibitor targeting the H /K -ATPase enzyme on gastric parietal cells It is designed to inhibit this proton pump thereby reducing gastric acid secretion Esomeprazole Magnesium exerts its biological activity primarily through selective and irreversible binding to H /K -ATPase inhibiting the final step in gastric acid secretion Based on these pharmacological properties Esomeprazole Magnesium holds research potential in studies of gastric acidity regulation gastroesophageal reflux disease (GERD) inflammation-related mucosal damage and investigations of pharmacokinetics and pharmacodynamics of proton pump inhibitors
Catalog No. 50-000-89188