Learn More
Selleck Chemical LLC NEXIUM (esomeprazole magnesium) 50mg 161973-10-0
SDP
Supplier:  Selleck Chemical LLC S174350MG
Encompass_Preferred
Esomeprazole Magnesium is a proton pump inhibitor to reduce gastric acid secretion through inhibition of H(+)/K(+)-ATPase in gastric parietal cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-529-0