Learn More
Medchemexpress LLC Esomeprazole magnesium (trihydrate) | 217087-09-7 | 99.6% | 767.17 | 1 G
SDP
Supplier:  Medchemexpress LLC HY170221G
Encompass_Preferred
Esomeprazole magnesium trihydrate is a potent and orally active H+, K+-ATPase inhibitor. It has potential for research into upper intestinal disorders and gastroesophageal reflux disease, and acts as an exosome inhibitor by blocking exosome release via the inhibition of V-H+-ATPases.
  • Potent and orally active H+, K+-ATPase inhibitor
  • Potential for research into upper intestinal disorders
  • Potential for gastroesophageal reflux disease research
  • Acts as an exosome inhibitor by blocking exosome release
  • Inhibits V-H+-ATPases
Catalog No. 50-003-84292