Learn More
Medchemexpress LLC Esomeprazole (potassium salt) | 161796-84-5 | 98.2% | 383.51 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17021B5MG
Encompass_Preferred
Esomeprazole potassium salt ((S)-Omeprazole potassium salt) is a potent and orally active proton pump inhibitor. It reduces acid secretion through inhibition of the H+, K+-ATPase in gastric parietal cells and has potential for symptomatic gastroesophageal reflux disease research.
  • Potent and orally active proton pump inhibitor
  • Reduces acid secretion by inhibiting H+, K+-ATPase
  • Potential for symptomatic gastroesophageal reflux disease research
  • Suppresses growth of triple-negative breast cancer cells in vitro
  • Inhibits progression of fibrosis in lungs in vivo
Catalog No. 50-003-85918