Learn More
Apexbio Technology LLC Esomeprazole Sodium 161796-78-7 50mg
SDP
Supplier:  Apexbio Technology LLC B138850
Encompass_Preferred
Esomeprazole Sodium the S-enantiomer of omeprazole functions pharmacologically as a proton pump inhibitor (PPI) modulating gastric acid secretion through selective inhibition of the H /K -ATPase enzyme in gastric parietal cells By irreversibly binding covalently to the sulfhydryl groups of this enzyme esomeprazole sodium suppresses gastric acid secretion thus serving as an experimental tool in biomedical research exploring acid-related gastrointestinal conditions including erosive esophagitis and gastroesophageal reflux disease (GERD) Reported IC50 values for inhibition of H /K -ATPase activity range approximately from 2 8 M highlighting usage in studies investigating gastric acid regulation and related signaling pathways at the molecular and cellular level
Catalog No. 50-000-62115