Learn More
Selleck Chemical LLC Esomeprazole sodium S2233-5mg
SDP
Supplier:  Selleck Chemical LLC S22335MG
Encompass_Preferred
Esomeprazole Sodium((S)-Omeprazole sodium (-)-Omeprazole sodium) is a sodium salt of esomeprazole that is a potent proton pump inhibitor with an IC50 of 0 076 mg/kg
Catalog No. 50-311-2792