Learn More
Medchemexpress LLC Esomeprazole (Standard) | 119141-88-7 | 99.77% | 345.42 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17021R5MG
Encompass_Preferred
Esomeprazole (Standard) is the analytical standard of Esomeprazole, a potent and orally active proton pump inhibitor that reduces acid secretion by inhibiting H+, K+-ATPase in gastric parietal cells. It has potential for research into symptomatic gastroesophageal reflux disease. This compound is intended for research and analytical applications, and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Analytical standard and reference standard supplied assay.
  • Used in HPLC, GC, and MS experiments.
  • For research purposes only.
Catalog No. 50-003-85074