Learn More
Med Vet International Esophagostomy Tube, 14Fr x 9", J0390, Each
Supplier:  Med Vet International JOR390
Encompass_Preferred
Esophagostomy Tube. Tubes for long term placement of esophagostomy tubes.• Soft silicone• Bio-compatible• Luer fitting• Retention cap always attached• Cut to desired length• Non-sterile• Packaged individually in chevron pouchEsophagostomy Tube. 14fr x 9”. For cats and small dogs.
Catalog No. 50-005-82741