Learn More
Cayman Chemical Estetrolhydrate 5mg
SDP
Supplier:  Cayman Chemical 182325 MG
Encompass_Preferred
An estrogen steroid hormone exclusively produced by fetal liver from estradiol and estriol during pregnancy; binds ERα with 4-fold higher affinity for over ERβ; exhibits estrogenic effects in bone, brain, vagina, and endometrium; acts as an estrogen antagonist in the presence of estradiol, preventing breast tumor growth
Catalog No. 50-296-7610