Learn More
Enzo Life Sciences 17beta-Estradiol high sensitivity ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900174
Encompass_Preferred
Highly sensitive ELISA, detecting as little as 14 pg/ml of 17β-Estradiol. The 17β-Estradiol (serum/plasma) EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Alternative Name: Estrogen. Sensitivity: 14.0 pg/ml (range 15.6 - 1,000 pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-946-890