Learn More
Cayman Chemical 17-Estradiol, 50-28-2, 1 g
SDP
Supplier:  Cayman Chemical 100063151
Encompass_Preferred
Molecular Formula C18H24O2, Purity ≥98%, Formula Weight 272.4, Synonyms: estra-1,3,5(10)-triene-3,17β-diol, β-Estradiol, Estradiol, 17β-Oestradiol, E2; The major estrogen secreted by the premenopausal ovary.
Catalog No. NC9986450