Learn More
Medchemexpress LLC Estradiol 3-methyl ether | 1035-77-4 | 99.9% | 286.41 | 5 MG
Supplier: Medchemexpress LLC HYW0085815MG
Estradiol 3-methyl ether, also known as 3-O-Methyl estradiol, is a synthetic estrogen. This compound has demonstrated the ability to disrupt microtubule networks in V79 cells with an EC50 of 9 μM, and it also inhibits cell proliferation. Furthermore, Estradiol 3-methyl ether has been observed to decrease serum cholesterol levels and contribute to body weight loss, indicating feminizing activity in an HFD murine model. It is intended for research use only.
- Functions as a synthetic version of the hormone estrogen.
- Can disrupt microtubule networks in V79 cells with an EC50 of 9 μM.
- Inhibits the proliferation of cells.
- Decreases serum cholesterol levels.
- Leads to an increase in body weight loss in HFD murine models.
- Exhibits feminizing activity in HFD murine models.