Learn More
Medchemexpress LLC HY-U00112 1mg Medchemexpress, Estradiol 3-sulfamate CAS:172377-52-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001121MG
Encompass_Preferred
Medchemexpress, HY-U00112 1mg Estradiol 3-sulfamate CAS:172377-52-5 Estradiol 3-sulfamate (BLE 00084; E2MATE; ES-J 995) is a potent, long-acting, and orally active steroid sulfatase inhibitor; inhibits estrone sulfatase with an IC 50 of 251 nM and a K i of 133 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4417