Learn More
Medchemexpress LLC Estradiol 3-sulfate 17-glucuronide potassium salt | 10392-35-5 | 98.3% | 604.75 g/mol | C24H30K2O11S | 500UG
SDP
Supplier:  Medchemexpress LLC HY123742500UG
Encompass_Preferred
Estradiol 3-sulfate 17-glucuronide (dipotassium salt) (CAS 10392-35-5) is a conjugated metabolite of estradiol used in biochemical and pharmacological research, including studies of steroid metabolism and transporter interactions. The compound is supplied as a research-grade dipotassium salt with reported solubility in common organic solvents; confirm lot-specific purity and storage conditions from the certificate of analysis and SDS.
Catalog No. 50-002-36799