Learn More
Cayman Chemical 17-Estradiol, 50-28-2, 5 g
SDP
Supplier:  Cayman Chemical 100063155
Encompass_Preferred
Molecular Formula C18H24O2, Purity ≥98%, Formula Weight 272.4, Synonyms: estra-1,3,5(10)-triene-3,17β-diol, β-Estradiol, Estradiol, 17β-Oestradiol, E2; The major estrogen secreted by the premenopausal ovary.
Catalog No. NC0294330
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.