Learn More
Apexbio Technology LLC Estradiol Benzoate 50-50-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19415.1
Encompass_Preferred
Estradiol benzoate (50-50-0) is a small-molecule agonist targeting estrogen receptors exhibiting high binding affinity for estrogen receptor alpha (ER ) It is designed to activate ER thereby modulating estrogen-mediated signaling pathways and hormonal regulation processes Estradiol benzoate exerts its biological activity primarily through ER agonism In in vitro assays involving human murine and chicken ER proteins estradiol benzoate demonstrates agonistic activity with an IC50 value ranging between 22 28 nM Based on these pharmacological properties estradiol benzoate holds research potential in studies of ER-mediated signaling endocrine-related diseases receptor-ligand binding analyses pharmacological screening and investigations of estrogen-dependent physiological and pathological conditions
Catalog No. 50-000-90018