Learn More
Medchemexpress LLC Estradiol/BSA | 95.0% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1615411MG
Encompass_Preferred
Estradiol/BSA is an antigen-adjuvant conjugate formed by coupling Estradiol with Bovine Serum Albumin (BSA). This conjugate is designed to enhance the production of antigen-specific antibodies in vaccine models by coupling the antigen with a protein adjuvant. It does not affect protein folding or disrupt major epitopes and can enhance cross-presentation and the generation of antigen-specific T cells. This product is for research use only.
  • Enhances production of antigen-specific antibodies
  • Used in vaccine models
  • Does not affect protein folding
  • Does not disrupt major epitopes
  • Enhances cross-presentation
  • Generates antigen-specific T cells
  • Antigen-adjuvant conjugate
Catalog No. 50-003-92619