Learn More
U.S. Pharmacopeia Estradiol cypionate, 313-06-4, MFCD00056558, 200 mg
Supplier:  U.S. Pharmacopeia 1252003
Encompass
Empirical Formula (Hill Notation): C26H36O3, Molecular Weight: 396.56, Synonym: β-Estradiol 17-cypionate, 1,3,5(10)-Estratriene-3,17β-diol 17-cyclopentylpropionate, 3,17β-Hydroxy-1,3,5(10)-estratriene 17-cyclopentylpropionate, Depoestradiol.
Catalog No. NC1444664