Learn More
Medchemexpress LLC Estradiol-d2-1 | 3188-46-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0141S65MG
Encompass_Preferred
Estradiol-d2-1 is a deuterium-labeled form of Estradiol, a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. It upregulates IL-6 expression via the estrogen receptor β (ERβ) pathway and is intended for research use only. Deuteration can impact pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, or LC-MS techniques.
  • Deuterium-labeled form of estradiol.
  • Upregulates IL-6 expression through the ERβ pathway.
Catalog No. 50-004-50857