Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol-d4 | 66789-03-5 | 99.12% | 5 MG
SDP

Supplier:  Medchemexpress LLC HYB0141S15MG

Encompass_Preferred

Estradiol-d4 is the deuterium-labeled form of Estradiol. Estradiol is a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. It upregulates IL-6 expression through the estrogen receptor β (ERβ) pathway. This compound can be utilized as a tracer or as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.

  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporated stable heavy isotopes for quantitation during drug development
  • Deuteration may influence pharmacokinetic and metabolic profiles
  • White to off-white solid
  • Molecular weight of 276.41
  • Chemical formula of C18H20D4O2

Catalog No. 50-004-50479


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.