Learn More
Medchemexpress LLC Estradiol dipropionate | 113-38-2 | MFCD00051146 | 99.5% | 384.51 g·mol⁻¹ | C24H32O4 | 10 MG

Supplier: Medchemexpress LLC HYB224510MG
Estradiol dipropionate is a steroidal estrogen ester used in biochemical and pharmacological research. It acts as an estrogen and progesterone agonist and is supplied in high-purity forms for use as a research reagent and analytical standard.
- High purity (99.5%) for reliable experimental results.
- Available as a solid powder and as a 10 mM solution in DMSO.
- Common package sizes include 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg.
- Molecular formula C24H32O4; molecular weight 384.51 g·mol⁻¹.
- CAS number 113-38-2 for unambiguous identification.
- Suitable for in vitro receptor assays and in vivo pharmacology studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.