Learn More
Cusabio Technology LLC Fish Estradiol(E2) ELISA kit
Supplier:  Cusabio Technology LLC CSBE13017FH
Encompass_Preferred
This Fish Estradiol(E2) ELISA kit was designed for the quantitative measurement of Fish Estradiol(E2) protein in serum, plasma. It is a Competitive ELISA kit, its detection range is 40 pg/ml-1000 pg/ml and the sensitivity is 40 pg/ml. One kit contains 96 wells.
Catalog No. NC2952861