Learn More
Medchemexpress LLC HY-B0672 500mg Medchemexpress, Estradiol valerianate CAS:979-32-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0672500MG
Encompass_Preferred
Medchemexpress, HY-B0672 500mg Estradiol valerianate CAS:979-32-8 Estradiol valerianate (β-estradiol 17-valerate) is a synthetic estrogen widely used in combination with other steroid hormones in hormone replacement therapy drugs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2638