Learn More
Medchemexpress LLC HY-N5060 500mg Medchemexpress, Estragole CAS:140-67-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN5060500MG
Encompass_Preferred
Medchemexpress, HY-N5060 500mg Estragole CAS:140-67-0 Estragole (4-Allylanisole), a relatively nontoxic volatile terpenoid ether, is a major component of the essential oil of many plants. Estragole dose-dependently blocks nerve excitability. Estragole displays anti-toxoplasma activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2941