Learn More
Medchemexpress LLC Estramustine phosphate | 4891-15-0 | 520.38 | 100 MG
Supplier: Medchemexpress LLC HY13627100MG
Estramustine phosphate, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. It depolymerizes microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. This action can interfere with mitosis, trigger cell death, and induce apoptosis, and can be used for the research of cancers like prostate cancer.
- Orally active antimicrotubule chemotherapy agent
- Depolymerizes microtubules
- Interferes with mitosis, triggers cell death, and induces apoptosis
- Used for prostate cancer research
- Suppresses PC3 cell growth
- Inhibits cell proliferation and tubulin cytoskeleton
- Inhibits TGF-β-induced cell migration