Learn More
Apexbio Technology LLC Estramustine phosphate sodium 52205-73-9 10mg
SDP
Supplier:  Apexbio Technology LLC A468610
Encompass_Preferred
Estramustine phosphate sodium (CAS 52205-73-9) is an orally bioavailable small molecule that functions as an antimicrotubule agent Structurally related to estradiol it interacts with microtubule-associated proteins (MAPs) promoting microtubule disassembly and disrupting mitotic spindle formation This action interferes with mitosis ultimately leading to apoptotic cell death Widely studied in prostate cancer models estramustine phosphate sodium serves as a valuable tool for investigating mechanisms of cell cycle regulation mitotic arrest and apoptosis in oncological and cellular biology research settings
Catalog No. 50-000-94986