Learn More
Medchemexpress LLC Estramustine phosphate sodium | 52205-73-9 | MFCD00866320 | 99.4% | C23H30Cl2NNa2O6P | 10MG
SDP
Supplier:  Medchemexpress LLC HY1362710MG
Encompass_Preferred
Estramustine phosphate sodium is an estradiol-derived, nitrogen-mustard conjugate used as an antimicrotubule research reagent. It depolymerizes microtubules, disrupts mitosis, and induces apoptosis, making it useful for in vitro studies of prostate and other hormone-related cancers.
  • Antimicrotubule activity causing mitotic arrest and apoptosis.
  • Water-soluble disodium phosphate salt suitable for aqueous assays.
  • High reported purity (≈99.4%) for reproducible results.
  • Available in small-mass packaging for screening and dose-response studies.
  • Characterized by standard identifiers for traceability.
  • Compatible with common cell-based and biochemical assays.
Catalog No. 50-002-61932