Learn More
Apexbio Technology LLC Estriol 50-27-1 500mg
SDP
Supplier:  Apexbio Technology LLC B1507500
Encompass_Preferred
Estriol (50-27-1) is a small-molecule estrogen receptor modulator targeting the G-protein coupled estrogen receptor (GPER) It is designed to antagonize GPER-dependent estrogen signaling thereby regulating estrogen-mediated cellular pathways independent of classical nuclear receptors Estriol exerts its biological activity primarily through antagonistic interaction with GPER In cell-based studies estriol demonstrates receptor-selective antagonistic activity in estrogen receptor-negative breast cancer cell lines Based on these pharmacological properties estriol holds research potential in oncology investigations involving hormone receptor-negative breast cancer and in the study of estrogen receptor-independent signaling pathways
Catalog No. 50-000-92046