Learn More
Selleck Chemical LLC Estriol S2466-50mg
SDP
Supplier:  Selleck Chemical LLC S246650MG
Encompass_Preferred
Estriol (NSC-12169) is an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells
Catalog No. 50-310-8858