Learn More
Apexbio Technology LLC Estropipate 7280-37-7 50mg
SDP
Supplier:  Apexbio Technology LLC B743850
Encompass_Preferred
Estropipate (7280-37-7) is a small-molecule agonist targeting estrogen receptors (ER) It is designed to activate ERs thereby modulating gene transcription and regulation of cell proliferation via estrogen receptor signaling pathways Estropipate exerts its biological activity primarily through metabolic conversion to active estrogens which bind to intracellular ERs and initiate receptor-mediated signaling Based on these pharmacological properties Estropipate holds research potential in the investigation of menopause-related physiological effects and estrogen-dependent bone metabolism and is frequently used in experimental models of osteoporosis and menopause-associated symptom management
Catalog No. 50-000-91698